Assay Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008960.2 | NP_032986.1 | 8 - 9 | 1895 | 78 | |
| GenBank mRNA | AK030750.1 | - | 4 - 5 | 658 | 78 | |
| AK076980.1 | - | 8 - 9 | 1853 | 78 | ||
| AK088717.1 | - | 8 - 9 | 1874 | 78 | ||
| AK138025.1 | - | 1 - 2 | 204 | 78 | ||
| AK148736.1 | - | 8 - 9 | 1730 | 78 | ||
| BC021445.1 | - | 8 - 9 | 1244 | 5038272 | 78 | |
| U92437.1 | - | 8 - 9 | 1975 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pten
Genomic Map