Assay Details
| Gene Symbol: Rasl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026878.1 | NP_081154.1 | 2 - 3 | 317 | 92 | |
| GenBank mRNA | BC008101.1 | - | 2 - 3 | 347 | 3156762 | 92 |
| BC083068.1 | - | 2 - 3 | 317 | 6308926 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasl11b
Genomic Map