Assay Details
| Gene Symbol: Aldh18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019698.2 | NP_062672.2 | 12 - 13 | 1492 | 89 | |
| NM_153554.2 | NP_705782.2 | 12 - 13 | 1486 | 89 | ||
| GenBank mRNA | AF056573.1 | - | 12 - 13 | 1397 | 89 | |
| AF056574.1 | - | 12 - 13 | 1391 | 89 | ||
| AK134148.1 | - | 12 - 13 | 1452 | 89 | ||
| AK147711.1 | - | 12 - 13 | 1412 | 89 | ||
| AK151072.1 | - | 12 - 13 | 1414 | 89 | ||
| AK153946.1 | - | 12 - 13 | 1413 | 89 | ||
| AK159610.1 | - | 12 - 13 | 1414 | 89 | ||
| AK166459.1 | - | 12 - 13 | 1378 | 89 | ||
| AK166494.1 | - | 12 - 13 | 1379 | 89 | ||
| AK168905.1 | - | 12 - 13 | 1412 | 89 | ||
| AK202353.1 | - | 232 | 89 | |||
| BC033427.1 | - | 12 - 13 | 1423 | 5009115 | 89 | |
| BC037699.1 | - | 12 - 13 | 1371 | 4188900 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh18a1
Genomic Map