Assay Details
| Gene Symbol: Aldh18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019698.2 | NP_062672.2 | 13 - 14 | 1717 | 113 | |
| NM_153554.2 | NP_705782.2 | 13 - 14 | 1711 | 113 | ||
| GenBank mRNA | AF056573.1 | - | 13 - 14 | 1622 | 113 | |
| AF056574.1 | - | 13 - 14 | 1616 | 113 | ||
| AK134148.1 | - | 13 - 14 | 1677 | 113 | ||
| AK147711.1 | - | 13 - 14 | 113 | |||
| AK151072.1 | - | 13 - 14 | 1639 | 113 | ||
| AK153946.1 | - | 13 - 14 | 1638 | 113 | ||
| AK159610.1 | - | 13 - 14 | 1639 | 113 | ||
| AK166459.1 | - | 13 - 14 | 1603 | 113 | ||
| AK166494.1 | - | 13 - 14 | 1604 | 113 | ||
| AK168905.1 | - | 13 - 14 | 1637 | 113 | ||
| BC033427.1 | - | 13 - 14 | 1648 | 5009115 | 113 | |
| BC037699.1 | - | 13 - 14 | 1596 | 4188900 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh18a1
Genomic Map