Assay Details
| Gene Symbol: Aldh18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019698.2 | NP_062672.2 | 7 - 8 | 956 | 70 | |
| GenBank mRNA | AF056573.1 | - | 7 - 8 | 861 | 70 | |
| AK134148.1 | - | 7 - 8 | 916 | 70 | ||
| AK147711.1 | - | 7 - 8 | 876 | 70 | ||
| AK151072.1 | - | 7 - 8 | 878 | 70 | ||
| AK153946.1 | - | 7 - 8 | 877 | 70 | ||
| AK159610.1 | - | 7 - 8 | 878 | 70 | ||
| AK166459.1 | - | 7 - 8 | 842 | 70 | ||
| AK166494.1 | - | 7 - 8 | 843 | 70 | ||
| AK196084.1 | - | 2 - 3 | 248 | 70 | ||
| BC033427.1 | - | 7 - 8 | 887 | 5009115 | 70 | |
| BC037699.1 | - | 7 - 8 | 835 | 4188900 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh18a1
Genomic Map