Assay Details
| Gene Symbol: Ash2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080793.2 | NP_001074262.1 | 2 - 3 | 181 | 91 | |
| NM_001286207.1 | NP_001273136.1 | 2 - 3 | 154 | 91 | ||
| NM_011791.3 | NP_035921.2 | 2 - 3 | 271 | 91 | ||
| GenBank mRNA | AB020983.2 | - | 2 - 3 | 253 | 91 | |
| AK087934.1 | - | 2 - 3 | 246 | 91 | ||
| AK132471.1 | - | 2 - 3 | 253 | 91 | ||
| AK145576.1 | - | 2 - 3 | 146 | 91 | ||
| AK145790.1 | - | 2 - 3 | 158 | 91 | ||
| AK145967.1 | - | 2 - 3 | 125 | 91 | ||
| AK146254.1 | - | 2 - 3 | 183 | 91 | ||
| AK146599.1 | - | 2 - 3 | 155 | 91 | ||
| AK146756.1 | - | 2 - 3 | 146 | 91 | ||
| AK146938.1 | - | 2 - 3 | 248 | 91 | ||
| BC012957.1 | - | 2 - 3 | 258 | 4211433 | 91 | |
| BY266419.1 | - | 2 - 3 | 264 | 91 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001080793.2,NM_001286207.1 transcripts.
This assay is in the 5'UTR region of the NM_001080793.2,NM_001286207.1 transcripts.
Set Membership: |
|

Ash2l
Genomic Map