Assay Details
| Gene Symbol: Kcnu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008432.3 | NP_032458.3 | 12 - 13 | 1329 | 103 | |
| GenBank mRNA | AF039213.2 | - | 12 - 13 | 1296 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnu1
Genomic Map