Assay Details
| Gene Symbol: Adar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038587.4 | NP_001033676.2 | 3 - 4 | 1745 | 121 | |
| NM_001146296.1 | NP_001139768.1 | 3 - 4 | 1737 | 121 | ||
| NM_019655.3 | NP_062629.3 | 3 - 4 | 1737 | 121 | ||
| GenBank mRNA | AF291875.1 | - | 2 - 3 | 212 | 121 | |
| AF291876.1 | - | 3 - 4 | 1644 | 121 | ||
| AF291877.1 | - | 2 - 3 | 212 | 121 | ||
| AK076413.1 | - | 3 - 4 | 1696 | 121 | ||
| AK089451.1 | - | 3 - 4 | 1708 | 121 | ||
| AK147614.1 | - | 3 - 4 | 1708 | 121 | ||
| AK156093.1 | - | 2 - 3 | 304 | 121 | ||
| AK172523.1 | - | 2 - 3 | 304 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adar
Genomic Map