Assay Details
| Gene Symbol: Fdps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253751.1 | NP_001240680.1 | 4 - 5 | 628 | 79 | |
| NM_134469.4 | NP_608219.1 | 3 - 4 | 412 | 79 | ||
| GenBank mRNA | AF309508.1 | - | 2 - 3 | 317 | 79 | |
| AK088601.1 | - | 3 - 4 | 332 | 79 | ||
| AK140881.1 | - | 3 - 4 | 404 | 79 | ||
| AK151742.1 | - | 3 - 4 | 341 | 79 | ||
| AK166026.1 | - | 3 - 4 | 331 | 79 | ||
| AK168008.1 | - | 3 - 4 | 331 | 79 | ||
| BC048497.1 | - | 3 - 4 | 390 | 6531586 | 79 | |
| BC087886.1 | - | 3 - 4 | 335 | 6595030 | 79 | |
| CT010365.1 | - | 2 - 3 | 284 | 79 | ||
More Information
Set Membership: |
|

Fdps
Genomic Map