Assay Details
| Gene Symbol: Mtx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161824.1 | NP_001155296.1 | 2 - 3 | 595 | 76 | |
| NM_013604.2 | NP_038632.2 | 2 - 3 | 595 | 76 | ||
| GenBank mRNA | AK086135.1 | - | 2 - 3 | 187 | 76 | |
| AK163982.1 | - | 2 - 3 | 163 | 76 | ||
| AK165923.1 | - | 2 - 3 | 168 | 76 | ||
| AX775987.1 | - | 2 - 3 | 167 | 76 | ||
| BB662550.1 | - | 2 - 3 | 187 | 76 | ||
| BC023071.1 | - | 2 - 3 | 166 | 5355724 | 76 | |
| L36962.1 | - | 2 - 3 | 165 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mtx1
Genomic Map