Assay Details
| Gene Symbol: Scamp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011886.2 | NP_036016.2 | 7 - 8 | 893 | 66 | |
| GenBank mRNA | AF005036.1 | - | 7 - 8 | 800 | 66 | |
| AF295403.1 | - | 7 - 8 | 798 | 66 | ||
| AK135335.1 | - | 7 - 8 | 877 | 66 | ||
| AK146204.1 | - | 7 - 8 | 895 | 66 | ||
| AK160549.1 | - | 7 - 8 | 878 | 66 | ||
| BC002021.1 | - | 7 - 8 | 891 | 3489293 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scamp3
Genomic Map