Assay Details
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163432.1 | NP_001156904.1 | 10 - 11 | 1437 | 71 | |
| NM_007712.3 | NP_031738.2 | 10 - 11 | 1440 | 71 | ||
| GenBank mRNA | AF033564.1 | - | 10 - 11 | 1162 | 71 | |
| AK134849.1 | - | 4 - 5 | 1681 | 71 | ||
| AK148377.1 | - | 8 - 9 | 2468 | 71 | ||
| AK161259.1 | - | 6 - 7 | 1612 | 71 | ||
| BC015080.1 | - | 10 - 11 | 1425 | 3995512 | 71 | |
| CN538691.1 | - | 5 - 6 | 602 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clk2
Genomic Map