Assay Details
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163432.1 | NP_001156904.1 | 11 - 12 | 1521 | 65 | |
| NM_007712.3 | NP_031738.2 | 11 - 12 | 1524 | 65 | ||
| GenBank mRNA | AF033564.1 | - | 11 - 12 | 1246 | 65 | |
| AK134849.1 | - | 5 - 6 | 1765 | 65 | ||
| AK161259.1 | - | 7 - 8 | 1696 | 65 | ||
| BC015080.1 | - | 11 - 12 | 1509 | 3995512 | 65 | |
| CD740936.1 | - | 2 - 3 | 65 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Clk2
Genomic Map