Assay Details
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163432.1 | NP_001156904.1 | 5 - 6 | 95 | ||
| NM_007712.3 | NP_031738.2 | 5 - 6 | 95 | |||
| GenBank mRNA | AK148377.1 | - | 3 - 4 | 1883 | 95 | |
| AK161259.1 | - | 1 - 2 | 95 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK148377.1 transcripts.
Set Membership: |
|

Clk2
Genomic Map