Assay Details
| Gene Symbol: Krtcap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025327.2 | NP_079603.1 | 4 - 5 | 364 | 75 | |
| GenBank mRNA | AK002479.1 | - | 4 - 5 | 365 | 75 | |
| AK003310.1 | - | 4 - 5 | 332 | 75 | ||
| BC031492.1 | - | 4 - 5 | 317 | 3485092 | 75 | |
| BC043030.1 | - | 4 - 5 | 836 | 5693076 | 75 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22318142 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22318141 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428505 | AK002479.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428505 | AK003310.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428505 | NM_025327.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428505 | BC031492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428505 | BC043030.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428505 | AK002479.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428505 | AK003310.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428505 | NM_025327.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428505 | BC031492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428505 | BC043030.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179596 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116004 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428505 | AK002479.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428505 | AK003310.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428505 | NM_025327.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428505 | BC031492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428505 | BC043030.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428505 | AK002479.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428505 | AK003310.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428505 | NM_025327.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428505 | BC031492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428505 | BC043030.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22318156 | - | - |
| Whole Mouse Genome G4122A | A_51_P428505 | AK002479.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428505 | AK003310.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428505 | NM_025327.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428505 | BC031492.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428505 | BC043030.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428505 | AK002479.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428505 | AK003310.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428505 | NM_025327.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428505 | BC031492.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428505 | BC043030.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127356 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119491 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02611865 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02611859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22318141 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22318142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22318107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03036760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03036764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02862878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03036752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02578708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03036766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02663410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02862877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03136830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03056884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03182269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22318139 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493487 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493489 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493485 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493488 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417059_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417058_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493484 | - | - |
| Mouse Genome 430 2.0 Array | 1417059_at | - | - |
| Mouse Genome 430 2.0 Array | 1417058_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417058_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417059_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66059_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66059_at | - | - |
| Mouse Expression Set 430 | 1417059_at | - | - |
| Mouse Expression Set 430 | 1417058_a_at | - | - |
| Mouse Expression Set 430 | 1417059_PM_at | - | - |
| Mouse Expression Set 430 | 1417058_PM_a_at | - | - |
| Murine Genome U74v2 Set | 97269_f_at | - | - |
| Murine Genome U74v2 Set | 97268_i_at | - | - |
| Murine Genome U74v2 Set | 97269_PM_f_at | - | - |
| Murine Genome U74v2 Set | 97268_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417058_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417059_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417059_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417058_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417059_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417058_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417058_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417059_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5220040 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5220040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005220040 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2605694 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2605694 | - | - |
| mouse-6 v1.1 (Illumina) | scl22978.7.1_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5220040 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22978.7.1_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5220040 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2605694 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2605694 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl22978.7.1_18-S | - | - |

Krtcap2
Genomic Map