Assay Details
| Gene Symbol: Pbxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146131.2 | NP_666243.1 | 9 - 10 | 984 | 80 | |
| GenBank mRNA | AK028754.1 | - | 9 - 10 | 945 | 80 | |
| AK028858.1 | - | 9 - 10 | 982 | 80 | ||
| AK159422.1 | - | 9 - 10 | 971 | 80 | ||
| AK159448.1 | - | 9 - 10 | 971 | 80 | ||
| AK159633.1 | - | 9 - 10 | 961 | 80 | ||
| AK160103.1 | - | 9 - 10 | 965 | 80 | ||
| AK170318.1 | - | 9 - 10 | 940 | 80 | ||
| BC026838.1 | - | 8 - 9 | 919 | 4460993 | 80 | |
| BC027771.1 | - | 9 - 10 | 954 | 5344900 | 80 | |
| BC034200.1 | - | 9 - 10 | 925 | 4986595 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pbxip1
Genomic Map