Assay Details
| Gene Symbol: Kcnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080466.2 | NP_536714.2 | 3 - 4 | 1753 | 79 | |
| GenBank mRNA | AK137738.1 | - | 3 - 4 | 1754 | 79 | |
| BC106769.1 | - | 3 - 4 | 1495 | 40039722 | 79 | |
| BC106770.1 | - | 3 - 4 | 1495 | 40039723 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnn3
Genomic Map