Assay Details
| Gene Symbol: Impad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177730.3 | NP_808398.1 | 2 - 3 | 749 | 65 | |
| GenBank mRNA | AK045511.1 | - | 2 - 3 | 799 | 65 | |
| BC048776.1 | - | 2 - 3 | 749 | 6439769 | 65 | |
| BC138209.1 | - | 2 - 3 | 606 | 8861228 | 65 | |
| BC145952.1 | - | 2 - 3 | 606 | 40131255 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Impad1
Genomic Map