Assay Details
| Gene Symbol: Txk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122754.2 | NP_001116226.1 | 3 - 4 | 257 | 120 | |
| NM_001289494.1 | NP_001276423.1 | 3 - 4 | 257 | 120 | ||
| NM_001289495.1 | NP_001276424.1 | 3 - 4 | 274 | 120 | ||
| NM_013698.3 | NP_038726.2 | 2 - 3 | 341 | 120 | ||
| GenBank mRNA | AK088209.1 | - | 3 - 4 | 253 | 120 | |
| AK138069.1 | - | 3 - 4 | 254 | 120 | ||
| AK138786.1 | - | 3 - 4 | 271 | 120 | ||
| BC120552.1 | - | 2 - 3 | 244 | 40129475 | 120 | |
| BC137710.1 | - | 2 - 3 | 246 | 8860729 | 120 | |
| D43963.1 | - | 2 - 3 | 341 | 120 | ||
| D83201.1 | - | 2 - 3 | 281 | 120 | ||
| D83202.1 | - | 2 - 3 | 444 | 120 | ||
| U16145.1 | - | 3 - 4 | 222 | 120 | ||
| U19607.1 | - | 3 - 4 | 257 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001289495.1 transcripts.
Set Membership: |
|

Txk
Genomic Map