Assay Details
| Gene Symbol: Tec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113460.2 | NP_001106931.1 | 11 - 12 | 1060 | 90 | |
| NM_001113461.2 | NP_001106932.1 | 10 - 11 | 994 | 90 | ||
| NM_001113464.2 | NP_001106935.1 | 10 - 11 | 973 | 90 | ||
| NM_013689.5 | NP_038717.2 | 10 - 11 | 953 | 90 | ||
| GenBank mRNA | AK139329.1 | - | 10 - 11 | 969 | 90 | |
| AK154457.1 | - | 11 - 12 | 1056 | 90 | ||
| AK168452.1 | - | 10 - 11 | 991 | 90 | ||
| AK168460.1 | - | 10 - 11 | 990 | 90 | ||
| AK170588.1 | - | 10 - 11 | 992 | 90 | ||
| BC018230.1 | - | 9 - 10 | 958 | 90 | ||
| BC037071.1 | - | 10 - 11 | 988 | 5029430 | 90 | |
| BC062884.1 | - | 6 - 7 | 536 | 5697182 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tec
Genomic Map