Assay Details
| Gene Symbol: Tec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113460.2 | NP_001106931.1 | 16 - 17 | 1717 | 83 | |
| NM_001113461.2 | NP_001106932.1 | 15 - 16 | 1651 | 83 | ||
| NM_001113464.2 | NP_001106935.1 | 15 - 16 | 1630 | 83 | ||
| NM_013689.5 | NP_038717.2 | 15 - 16 | 1610 | 83 | ||
| GenBank mRNA | AK139329.1 | - | 15 - 16 | 1626 | 83 | |
| AK154457.1 | - | 16 - 17 | 1713 | 83 | ||
| AK168452.1 | - | 15 - 16 | 1648 | 83 | ||
| AK168460.1 | - | 15 - 16 | 1647 | 83 | ||
| AK170588.1 | - | 15 - 16 | 1649 | 83 | ||
| BC018230.1 | - | 14 - 15 | 1613 | 83 | ||
| BC037071.1 | - | 15 - 16 | 1645 | 5029430 | 83 | |
| BC062884.1 | - | 11 - 12 | 1193 | 5697182 | 83 | |
| M33427.1 | - | 1 - 2 | 77 | 83 | ||
| S53716.1 | - | 15 - 16 | 1641 | 83 | ||
| X55663.1 | - | 15 - 16 | 1645 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tec
Genomic Map