Assay Details
| Gene Symbol: Tec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113460.2 | NP_001106931.1 | 7 - 8 | 690 | 97 | |
| NM_001113461.2 | NP_001106932.1 | 7 - 8 | 690 | 97 | ||
| NM_001113464.2 | NP_001106935.1 | 6 - 7 | 603 | 97 | ||
| NM_013689.5 | NP_038717.2 | 7 - 8 | 649 | 97 | ||
| GenBank mRNA | AK133418.1 | - | 7 - 8 | 650 | 97 | |
| AK139329.1 | - | 6 - 7 | 599 | 97 | ||
| AK168452.1 | - | 7 - 8 | 687 | 97 | ||
| AK168460.1 | - | 7 - 8 | 686 | 97 | ||
| AK170588.1 | - | 7 - 8 | 688 | 97 | ||
| BC018230.1 | - | 6 - 7 | 654 | 97 | ||
| BC037071.1 | - | 7 - 8 | 684 | 5029430 | 97 | |
| BC062884.1 | - | 2 - 3 | 166 | 5697182 | 97 | |
| S53716.1 | - | 6 - 7 | 614 | 97 | ||
| X55663.1 | - | 7 - 8 | 684 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062884.1 transcripts.
Set Membership: |
|

Tec
Genomic Map