Assay Details
| Gene Symbol: Tsacc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029801.2 | NP_084077.1 | 3 - 4 | 255 | 67 | |
| GenBank mRNA | AK006195.1 | - | 3 - 4 | 256 | 67 | |
| BC107298.2 | - | 3 - 4 | 221 | 40053153 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22312953 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22312952 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22312954 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131653 | BC107298.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131653 | NM_029801.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131653 | AK006195.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131653 | BC107298.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131653 | NM_029801.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P131653 | AK006195.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482815 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255789 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628757 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267045 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778098 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99130 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131948 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154939 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131653 | BC107298.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131653 | NM_029801.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131653 | AK006195.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131653 | BC107298.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131653 | NM_029801.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P131653 | AK006195.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131948 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154939 | - | - |
| Whole Mouse Genome G4122A | A_52_P610541 | BC107298.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P610541 | NM_029801.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P610541 | AK006195.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131653 | BC107298.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131653 | NM_029801.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131653 | AK006195.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131653 | BC107298.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131653 | NM_029801.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131653 | AK006195.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131948 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154939 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03161982 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03161980 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03161977 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03097039 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22312953 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22312954 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22312952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22312933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02923341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02981815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02814727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03031754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03193787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02781756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03161980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02679370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02366314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22312946 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499345 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499347 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499346 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499343 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499344 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429961_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499342 | - | - |
| Mouse Genome 430 2.0 Array | 1429961_at | - | - |
| Mouse Genome 430 2.0 Array | 1429961_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76927_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76927_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300017440.mm.1 | NM_029801.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017440.mm.1 | NM_029801.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017440.mm.1 | TC0300002305.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429961_at | - | - |
| Mouse Expression Set 430 | 1429961_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429961_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429961_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 542023 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 540041 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232146 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232146 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1232146 | - | - |

Tsacc
Genomic Map