| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400002555.mm.1 |
NM_001166531.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002555.mm.1 |
NM_001166531.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002555.mm.1 |
NM_001166532.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002555.mm.1 |
NM_001166532.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002555.mm.1 |
NM_019460.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002555.mm.1 |
NM_019460.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002555.mm.1 |
TC1400000310.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400002554.mm.1 |
NM_001166531.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002554.mm.1 |
NM_001166531.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002554.mm.1 |
NM_001166532.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002554.mm.1 |
NM_001166532.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002554.mm.1 |
NM_019460.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002554.mm.1 |
NM_019460.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002554.mm.1 |
TC1400000310.mm.1 |
MTA TC |