Assay Details
| Gene Symbol: Sfmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166531.1 | NP_001160003.1 | 16 - 17 | 1885 | 73 | |
| NM_001166532.1 | NP_001160004.1 | 15 - 16 | 1979 | 73 | ||
| NM_019460.2 | NP_062333.1 | 15 - 16 | 1836 | 73 | ||
| GenBank mRNA | AB032165.1 | - | 14 - 15 | 1681 | 73 | |
| AK142330.1 | - | 15 - 16 | 1864 | 73 | ||
| AK154826.1 | - | 15 - 16 | 1981 | 73 | ||
| AK160573.1 | - | 16 - 17 | 1885 | 73 | ||
| BC018217.1 | - | 15 - 16 | 1847 | 73 | ||
| BC036972.1 | - | 15 - 16 | 1906 | 73 | ||
| BC066188.1 | - | 15 - 16 | 1789 | 30476111 | 73 | |
More Information
Set Membership: |
|

Sfmbt1
Genomic Map