Assay Details
| Gene Symbol: Sfmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166531.1 | NP_001160003.1 | 21 - 22 | 2732 | 72 | |
| NM_001166532.1 | NP_001160004.1 | 20 - 21 | 2826 | 72 | ||
| NM_019460.2 | NP_062333.1 | 20 - 21 | 2683 | 72 | ||
| GenBank mRNA | AB032165.1 | - | 19 - 20 | 2528 | 72 | |
More Information
Set Membership: |
|

Sfmbt1
Genomic Map