Assay Details
| Gene Symbol: Zcchc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019621.1 | - | 1 - 2 | 143 | 90 | |
| AK049511.1 | - | 1 - 2 | 150 | 90 | ||
| AK138997.1 | - | 1 - 2 | 162 | 90 | ||
More Information
Set Membership: |
|

Zcchc4
Genomic Map