Assay Details
| Gene Symbol: Zcchc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019621.1 | - | 2 - 3 | 263 | 93 | |
| AK049511.1 | - | 2 - 3 | 270 | 93 | ||
| AK138997.1 | - | 2 - 3 | 282 | 93 | ||
| AK143805.1 | - | 2 - 3 | 231 | 93 | ||
More Information
Set Membership: |
|

Zcchc4
Genomic Map