Assay Details
| Gene Symbol: Zcchc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019621.1 | - | 4 - 5 | 621 | 62 | |
| AK049511.1 | - | 4 - 5 | 628 | 62 | ||
| AK138997.1 | - | 4 - 5 | 640 | 62 | ||
| AK143805.1 | - | 4 - 5 | 589 | 62 | ||
| BC107392.2 | - | 546 | 40057902 | 62 | ||
| BC107393.2 | - | 546 | 40057909 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107392.2, BC107393.2 transcripts.
Set Membership: |
|

Zcchc4
Genomic Map