Assay Details
| Gene Symbol: Dhx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007839.3 | NP_031865.2 | 12 - 13 | 2298 | 107 | |
| NR_104311.1 | 12 - 13 | 2290 | 107 | |||
| GenBank mRNA | AF017153.1 | - | 12 - 13 | 2146 | 107 | |
| AK145983.1 | - | 12 - 13 | 2248 | 107 | ||
| AK146441.1 | - | 12 - 13 | 2245 | 107 | ||
| AK151757.1 | - | 12 - 13 | 2261 | 107 | ||
| AK167927.1 | - | 12 - 13 | 2169 | 107 | ||
| AK168835.1 | - | 12 - 13 | 2266 | 107 | ||
| AK169174.1 | - | 12 - 13 | 2266 | 107 | ||
| BC003745.1 | - | 12 - 13 | 2241 | 107 | ||
| BC144759.1 | - | 12 - 13 | 2121 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhx15
Genomic Map