Assay Details
| Gene Symbol: Hoga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026152.1 | NP_080428.1 | 4 - 5 | 656 | 73 | |
| GenBank mRNA | AK002457.1 | - | 4 - 5 | 656 | 73 | |
| AK010857.1 | - | 4 - 5 | 616 | 73 | ||
| BC016430.1 | - | 4 - 5 | 625 | 4235003 | 73 | |
More Information
Set Membership: |
|

Hoga1
Genomic Map