Assay Details
| Gene Symbol: Trmt44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030208.3 | NP_084484.2 | 3 - 4 | 847 | 93 | |
| GenBank mRNA | AK019101.1 | - | 3 - 4 | 838 | 93 | |
| AK046957.1 | - | 3 - 4 | 849 | 93 | ||
| AK132402.1 | - | 3 - 4 | 848 | 93 | ||
| BC152332.1 | - | 3 - 4 | 842 | 40141614 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt44
Genomic Map