Assay Details
| Gene Symbol: Grk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080743.1 | NP_001074212.1 | 4 - 5 | 913 | 161 | |
| NM_019497.2 | NP_062370.2 | 4 - 5 | 913 | 161 | ||
| GenBank mRNA | AK020792.1 | - | 4 - 5 | 913 | 161 | |
| AK132685.1 | - | 4 - 5 | 914 | 161 | ||
| BC150691.1 | - | 4 - 5 | 437 | 9087602 | 161 | |
More Information
Set Membership: |
|

Grk4
Genomic Map