Assay Details
| Gene Symbol: Grk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019497.2 | NP_062370.2 | 9 - 10 | 1495 | 94 | |
| GenBank mRNA | AF040745.1 | - | 9 - 10 | 921 | 94 | |
| AK132685.1 | - | 9 - 10 | 1496 | 94 | ||
| BC150691.1 | - | 9 - 10 | 1019 | 9087602 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grk4
Genomic Map