Assay Details
| Gene Symbol: Cpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153107.2 | NP_694747.2 | 7 - 8 | 1331 | 89 | |
| GenBank mRNA | AF356844.1 | - | 7 - 8 | 1240 | 89 | |
| BX518878.1 | - | 1 - 2 | 158 | 89 | ||
More Information
Set Membership: |
|

Cpz
Genomic Map