Assay Details
| Gene Symbol: Nop14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029278.2 | NP_083554.2 | 15 - 16 | 2312 | 78 | |
| GenBank mRNA | AK148605.1 | - | 15 - 16 | 2270 | 78 | |
| AK161728.1 | - | 15 - 16 | 2250 | 78 | ||
| BC043043.1 | - | 15 - 16 | 2255 | 5701430 | 78 | |
More Information
Set Membership: |
|

Nop14
Genomic Map