Assay Details
| Gene Symbol: Nop14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029278.2 | NP_083554.2 | 3 - 4 | 574 | 73 | |
| GenBank mRNA | AK011648.1 | - | 3 - 4 | 550 | 73 | |
| AK078605.1 | - | 3 - 4 | 560 | 73 | ||
| AK079609.1 | - | 3 - 4 | 576 | 73 | ||
| AK148605.1 | - | 3 - 4 | 533 | 73 | ||
| AK161728.1 | - | 3 - 4 | 512 | 73 | ||
| BC024998.1 | - | 3 - 4 | 495 | 5012070 | 73 | |
| BC043043.1 | - | 3 - 4 | 517 | 5701430 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nop14
Genomic Map