| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500003832.mm.1 |
TC0500000387.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003832.mm.1 |
NM_001163512.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003832.mm.1 |
NM_001163512.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003832.mm.1 |
NM_173402.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003832.mm.1 |
NM_173402.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003833.mm.1 |
TC0500000387.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003833.mm.1 |
NM_001163512.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003833.mm.1 |
NM_001163512.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003833.mm.1 |
NM_173402.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003833.mm.1 |
NM_173402.2 |
Refseq |