Assay Details
| Gene Symbol: Acox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030721.2 | NP_109646.2 | 8 - 9 | 963 | 79 | |
| GenBank mRNA | AJ278430.1 | - | 8 - 9 | 996 | 79 | |
| AK028818.1 | - | 8 - 9 | 1384 | 79 | ||
| AK053960.1 | - | 8 - 9 | 960 | 79 | ||
| AK128922.1 | - | 8 - 9 | 923 | 79 | ||
| AK154112.1 | - | 7 - 8 | 1054 | 79 | ||
| AK171598.1 | - | 946 | 79 | |||
| BC044725.2 | - | 8 - 9 | 983 | 5374281 | 79 | |
| BC055019.1 | - | 8 - 9 | 963 | 4162223 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acox3
Genomic Map