Assay Details
| Gene Symbol: Rfc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027009.2 | NP_081285.1 | 3 - 4 | 350 | 69 | |
| GenBank mRNA | AK013095.1 | - | 3 - 4 | 338 | 69 | |
| AK042393.1 | - | 3 - 4 | 351 | 69 | ||
| AK165891.1 | - | 3 - 4 | 345 | 69 | ||
| AK167047.1 | - | 3 - 4 | 351 | 69 | ||
| BC026795.1 | - | 3 - 4 | 321 | 4015134 | 69 | |
More Information
Set Membership: |
|

Rfc3
Genomic Map