Assay Details
| Gene Symbol: Rfc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027009.2 | NP_081285.1 | 8 - 9 | 939 | 103 | |
| GenBank mRNA | AK145126.1 | - | 8 - 9 | 966 | 103 | |
| AK167047.1 | - | 8 - 9 | 940 | 103 | ||
| BC026795.1 | - | 8 - 9 | 910 | 4015134 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479434 | BC026795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479434 | AK042393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479434 | AK145126.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479434 | AK165891.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479434 | AK167047.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479434 | BC026795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479434 | AK042393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479434 | AK145126.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479434 | AK165891.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479434 | AK167047.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 252109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396299 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955636 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136258 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171305 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479434 | BC026795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479434 | AK042393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479434 | AK145126.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479434 | AK165891.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479434 | AK167047.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479434 | BC026795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479434 | AK042393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479434 | AK145126.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479434 | AK165891.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479434 | AK167047.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171305 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136258 | - | - |
| Whole Mouse Genome G4122A | A_51_P479434 | BC026795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479434 | AK042393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479434 | AK145126.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479434 | AK165891.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479434 | AK167047.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479434 | BC026795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479434 | AK042393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479434 | AK145126.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479434 | AK165891.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479434 | AK167047.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136258 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24197077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24197097 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535987 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535980 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535988 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535986 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432538_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423700_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535979 | - | - |
| Mouse Genome 430 2.0 Array | 1423700_at | - | - |
| Mouse Genome 430 2.0 Array | 1432538_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432538_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423700_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69263_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69263_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500033789.mm.1 | TC0500003705.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500033789.mm.1 | NM_027009.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500033789.mm.1 | NM_027009.2 | Refseq |
| Mouse Expression Set 430 | 1423700_PM_at | - | - |
| Mouse Expression Set 430 | 1423700_at | - | - |
| Mouse Expression Set 430 | 1432538_PM_a_at | - | - |
| Mouse Expression Set 430 | 1432538_a_at | - | - |
| Murine Genome U74v2 Set | 160072_at | - | - |
| Murine Genome U74v2 Set | 160072_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423700_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432538_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423700_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432538_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423700_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432538_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432538_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423700_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 760949 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106770672 | - | - |
| MouseWG-6 v2 Expression BeadChip | 1980600 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106770672 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257617 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257617 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2543834 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106770672 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106770672 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2726047 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2543834 | - | - |

Rfc3
Genomic Map