Assay Details
| Gene Symbol: Chrnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173212.4 | NP_775304.1 | 2 - 3 | 476 | 100 | |
| GenBank mRNA | AF325350.1 | - | 1 - 2 | 84 | 100 | |
| AF467896.1 | - | 2 - 3 | 239 | 100 | ||
| AK030464.1 | - | 2 - 3 | 477 | 100 | ||
| AK133689.1 | - | 2 - 3 | 420 | 100 | ||
| AY574268.1 | - | 2 - 3 | 218 | 100 | ||
| BC058193.1 | - | 2 - 3 | 239 | 5357762 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chrnb3
Genomic Map