Assay Details
| Gene Symbol: Chrnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027454.4 | NP_081730.1 | 3 - 4 | 592 | 72 | |
| NM_173212.4 | NP_775304.1 | 4 - 5 | 637 | 72 | ||
| GenBank mRNA | AF325350.1 | - | 3 - 4 | 245 | 72 | |
| AF467896.1 | - | 4 - 5 | 400 | 72 | ||
| AK017571.1 | - | 3 - 4 | 522 | 72 | ||
| AK030464.1 | - | 4 - 5 | 638 | 72 | ||
| AK133689.1 | - | 4 - 5 | 581 | 72 | ||
| AY574268.1 | - | 4 - 5 | 379 | 72 | ||
| BC058193.1 | - | 4 - 5 | 400 | 5357762 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chrnb3
Genomic Map