Assay Details
| Gene Symbol: Ets2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011809.3 | NP_035939.3 | 5 - 6 | 746 | 90 | |
| GenBank mRNA | AK014602.2 | - | 5 - 6 | 668 | 90 | |
| AK030537.1 | - | 1 - 2 | 48 | 90 | ||
| AK038348.1 | - | 5 - 6 | 733 | 90 | ||
| AK128933.1 | - | 5 - 6 | 698 | 90 | ||
| AK143687.1 | - | 5 - 6 | 698 | 90 | ||
| AK144974.1 | - | 5 - 6 | 697 | 90 | ||
| AK145852.1 | - | 5 - 6 | 582 | 90 | ||
| AK150175.1 | - | 5 - 6 | 676 | 90 | ||
| AK151315.1 | - | 5 - 6 | 697 | 90 | ||
| AK152750.1 | - | 5 - 6 | 700 | 90 | ||
| AK153339.1 | - | 5 - 6 | 698 | 90 | ||
| AK153478.1 | - | 5 - 6 | 698 | 90 | ||
| BC005486.1 | - | 5 - 6 | 667 | 3486021 | 90 | |
| BC005504.1 | - | 5 - 6 | 607 | 3490813 | 90 | |
| CT010235.1 | - | 4 - 5 | 508 | 90 | ||
| J04103.1 | - | 5 - 6 | 645 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ets2
Genomic Map