Assay Details
| Gene Symbol: Ets2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011809.3 | NP_035939.3 | 6 - 7 | 830 | 66 | |
| GenBank mRNA | AK014602.2 | - | 6 - 7 | 752 | 66 | |
| AK030537.1 | - | 2 - 3 | 132 | 66 | ||
| AK038348.1 | - | 6 - 7 | 817 | 66 | ||
| AK128933.1 | - | 6 - 7 | 782 | 66 | ||
| AK143687.1 | - | 6 - 7 | 782 | 66 | ||
| AK144974.1 | - | 6 - 7 | 781 | 66 | ||
| AK145852.1 | - | 6 - 7 | 666 | 66 | ||
| AK151315.1 | - | 6 - 7 | 781 | 66 | ||
| AK152750.1 | - | 6 - 7 | 784 | 66 | ||
| AK153339.1 | - | 6 - 7 | 782 | 66 | ||
| AK153478.1 | - | 6 - 7 | 782 | 66 | ||
| BC005504.1 | - | 6 - 7 | 691 | 3490813 | 66 | |
| J04103.1 | - | 6 - 7 | 729 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ets2
Genomic Map