Assay Details
| Gene Symbol: Nek4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF099067.1 | - | 12 - 13 | 2085 | 113 | |
| AJ223071.1 | - | 12 - 13 | 2175 | 113 | ||
| AK078809.1 | - | 11 - 12 | 2052 | 113 | ||
| AK132803.1 | - | 11 - 12 | 1969 | 113 | ||
| AK149147.1 | - | 3 - 4 | 253 | 113 | ||
| AK149584.1 | - | 11 - 12 | 1969 | 113 | ||
| BC057939.1 | - | 12 - 13 | 2087 | 5325558 | 113 | |
| Y09234.1 | - | 11 - 12 | 2031 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nek4
Genomic Map