Assay Details
| Gene Symbol: Gnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153547.6 | NP_705775.2 | 4 - 5 | 504 | 74 | |
| NR_104298.1 | 4 - 5 | 504 | 74 | |||
| GenBank mRNA | AK077523.1 | - | 4 - 5 | 435 | 74 | |
| AK167094.1 | - | 2 - 3 | 1923 | 74 | ||
| AK167183.1 | - | 4 - 5 | 388 | 74 | ||
| AY181025.1 | - | 4 - 5 | 362 | 74 | ||
| AY185498.1 | - | 4 - 5 | 362 | 74 | ||
| BC018560.1 | - | 4 - 5 | 391 | 74 | ||
| BC037996.1 | - | 4 - 5 | 364 | 5052858 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnl3
Genomic Map