Assay Details
| Gene Symbol: Gnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153547.6 | NP_705775.2 | 5 - 6 | 595 | 79 | |
| GenBank mRNA | AK077523.1 | - | 5 - 6 | 526 | 79 | |
| AK167183.1 | - | 5 - 6 | 479 | 79 | ||
| AY181025.1 | - | 5 - 6 | 453 | 79 | ||
| AY185498.1 | - | 5 - 6 | 453 | 79 | ||
| BC037996.1 | - | 5 - 6 | 455 | 5052858 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnl3
Genomic Map