Assay Details
| Gene Symbol: Gnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153547.6 | NP_705775.2 | 6 - 7 | 725 | 109 | |
| NR_104298.1 | 6 - 7 | 677 | 109 | |||
| GenBank mRNA | AK077523.1 | - | 6 - 7 | 656 | 109 | |
| AK167094.1 | - | 4 - 5 | 2158 | 109 | ||
| AK167183.1 | - | 6 - 7 | 609 | 109 | ||
| AY181025.1 | - | 6 - 7 | 583 | 109 | ||
| AY185498.1 | - | 6 - 7 | 583 | 109 | ||
| BC018560.1 | - | 6 - 7 | 564 | 109 | ||
| BC037996.1 | - | 6 - 7 | 585 | 5052858 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnl3
Genomic Map