Assay Details
| Gene Symbol: Gm14743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126321.2 | NP_001119793.1 | 2 - 3 | 217 | 121 | |
More Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			Set Membership: | 
		   
					
						
							 | 
				

Gm14743
Genomic Map